Anti-NEB Mouse Monoclonal Antibody [clone: 10K287]
Catalog # USBIN0570-74A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Nebulin
- Antigen symbol:NEB
- Clonality:Monoclonal
- Clone:10K287
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Frog,Chicken,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:1.2% sodium acetate, 2mg BSA, 0,01mg sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-2?g/ml
Type: Primary
Antigen: NEB (nebulin)
Clonality: monoclonal
Clone: 10K287
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Frog, Human, Mouse, Rat