Anti-Glycophorin Mouse monoclonal antibody [clone: A63-B/C2]
USBIG8177-01
- Antibody type:Primary
- Antigen name:Glycophorin
- Clonality:Monoclonal
- Clone:A63-B/C2
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Glycophorin
Clonality: monoclonal
Clone: A63-B/C2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human