Ordreindtastning
Denmark
ContactUsLinkComponent
Anti-Connexin 31.1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anti-Connexin 31.1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
  USBIC7853-61F-FITC
undefined
Anti-Connexin 31.1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
  USBIC7853-61F-FITC

 

  • Antistof type:
    Primary
  • Antigen name:
    Connexin 31.1
  • Clonality:
    Polyclonal
  • Konjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Vært:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Epitop:
    internal
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Opbevaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    purified by Protein G chromatography
  • Størrelse:
    200 µl
  • Pakn:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: Connexin 31.1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human