Anti-Complement C2 Rabbit Polyclonal Antibody
Catalog # USBIC7850-09C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Complement C2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Format:Serum
- Form:lyophilized powder
- Storage buffer:PBS,EDTA. 1ml sterile ddH2O. Dilute further
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: Complement C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human