Order Entry
Denmark
ContactUsLinkComponent
Anti-Complement C2 Mouse Monoclonal Antibody [clone: 6F61]
Anti-Complement C2 Mouse Monoclonal Antibody [clone: 6F61]
  USBIC7850-09A
Anti-Complement C2 Mouse Monoclonal Antibody [clone: 6F61]
  USBIC7850-09A

 

  • Antibody type:
    Primary
  • Antigen name:
    Complement C2
  • Clonality:
    Monoclonal
  • Clone:
    6F61
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 15mM sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A/G chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Recommended dilution: ELISA: 1:4000; Western Blot: 1:500

Type: Primary
Antigen: Complement C2
Clonality: Monoclonal
Clone: 6F61
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human