Anti-Chloramphenicol Mouse Monoclonal Antibody [clone: 10i2]
Catalog # USBIC4350-01H
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Chloramphenicol
- Clonality:Monoclonal
- Clone:10i2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Chloramphenicol
Clonality: Monoclonal
Clone: 10i2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: