Anti-IL2RA Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5F29]
Catalog # USBIC2275-94A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 2 Receptor Alpha
- Antigen symbol:IL2RA
- Clonality:Monoclonal
- Clone:5F29
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 Tests
- Pk:50 Tests
Specifications
About this item
Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: Monoclonal
Clone: 5F29
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human