Anti-CD4, Low Endotoxin Rat Monoclonal Antibody [clone: 10A3]
Catalog # USBIC2255-05C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CD4
- Clonality:Monoclonal
- Clone:10A3
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rat
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:25-1:200
Type: Primary
Antigen: CD4, Low Endotoxin
Clonality: Monoclonal
Clone: 10A3
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse