Anti-Blue Fluorescence Protein, Enhanced Mouse Monoclonal Antibody [clone: 3A6]
USBIB2174-93
- Antibody type:Tag
- Antigen name:Blue Fluorescence Protein, Enhanced
- Clonality:Monoclonal
- Clone:3A6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:10mMammonium bicarbonate.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Tag
Antigen: Blue Fluorescence Protein, Enhanced
Clonality: monoclonal
Clone: 3A6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: