Anti-BAD Mouse monoclonal antibody unconjugated [clone: 10i30]
Catalog # USBIB0003-47H
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:BCL2-associated agonist of cell death
- Antigen symbol:BAD
- Clonality:Monoclonal
- Clone:10i30
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Modification:phospho-Ser155
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Type: Primary
Antigen: BAD (BCL2-associated agonist of cell death)
Clonality: monoclonal
Clone: 10i30
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse