Anti-PRKAG3 Goat Polyclonal Antibody
Catalog # USBIA1475-08F
Leverandør: US Biological
Specifikationer
- Antibody type:Primary
- Antigen name:Protein Kinase AMP-Activated Non-Catalytic Subunit Gamma 3
- Antigen symbol:PRKAG3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Form:liquid
- Storage buffer:Tris, pH 7,2, 0,5% BSA, 0,02% sodium azide.
- Sequence:C-KLGWDDELRKPGAQ
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifikationer
Om denne vare
Recommended dilution: ELISA: 1:8000
Type: Primary
Antigen: PRKAG3 (protein kinase, AMP-activated, gamma 3 non-catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human