Order Entry
Denmark
ContactUsLinkComponent
Anti-AMPK1, 2 Rabbit Polyclonal Antibody
Anti-AMPK1, 2 Rabbit Polyclonal Antibody
Catalog # USBIA1475-08E3
Supplier:  US Biological
Anti-AMPK1, 2 Rabbit Polyclonal Antibody
Catalog # USBIA1475-08E3
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase, AMP-Activated, Alpha 1/2 Catalytic Subunit
  • Antigen symbol:
    AMPK1/2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Form:
    liquid
  • Antigen synonyms:
    PRKAA1/2
  • Storage buffer:
    PBS(without Mg2+ and Ca2+), pH 7,4, 150mMsodium chloride, 0,02% sodium azide, 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: AMPK1, 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat