Order Entry
Denmark
ContactUsLinkComponent
Anti-PRKAG1 Rabbit Polyclonal Antibody
Anti-PRKAG1 Rabbit Polyclonal Antibody
Catalog # USBIA1475-07
Supplier:  US Biological
Anti-PRKAG1 Rabbit Polyclonal Antibody
Catalog # USBIA1475-07
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase AMP-Activated Non-Catalytic Subunit Gamma 1
  • Antigen symbol:
    PRKAG1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    10mMsodium HEPES, pH 7,5, 150mM NaCl, 100ug/ml BSA, 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A and peptide chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: PRKAG1 (protein kinase, AMP-activated, gamma 1 non-catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse