Anti-PRKAB1 Rabbit Polyclonal Antibody (Biotin)
USBIA1475-03C-BIOT
- Antibody type:Primary
- Antigen name:Protein Kinase AMP-Activated Non-Catalytic Subunit Beta 1
- Antigen symbol:PRKAB1
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: PRKAB1 (protein kinase, AMP-activated, beta 1 non-catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse