Anti-PRKAA1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
USBIA1475-01Q1-APC
- Antibody type:Primary
- Antigen name:Protein kinase, AMP-activated, alpha 1 catalytic subunit
- Antigen symbol:PRKAA1
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Do not freeze!
Type: Primary
Antigen: PRKAA1 (protein kinase, AMP-activated, alpha 1 catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human