Anti-AKR7A2 Rabbit Polyclonal Antibody
USBIA1334-76A
- Antibody type:Primary
- Antigen name:Aldo-Keto Reductase family 7, member A2 (aflatoxin aldehyde reductase)
- Antigen symbol:AKR7A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Type: Primary
Antigen: AKR7A2 (aldo-keto reductase family 7, member A2 (aflatoxin aldehyde reductase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse