Anti-Aggrecan ARGxx Mouse Monoclonal Antibody [clone: 8J209]
USBIA1059-53U
- Antibody type:Primary
- Antigen name:Aggrecan ARGxx
- Clonality:Monoclonal
- Clone:8J209
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Pig,Rabbit,Horse,Sheep,Human,Bovine,Guinea pig,Dog,Rat,Cat
- Western blot:Yes
- Environmentally Preferable:
- Format:Tissue culture supernatant
- Form:liquid
- Storage buffer:0,2M Tris/HCl pH 7,4, 01-10% fetal calf serum, 40% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:100 µG
Recommended dilution: Western Blot: 1:50
Type: Primary
Antigen: Aggrecan ARGxx
Clonality: monoclonal
Clone: 8J209
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine, Cat, Dog, Guinea pig, Horse, Human, Pig, Rabbit, Rat, Sheep