Anti-AK1 Mouse Monoclonal Antibody [clone: 3A6-1F5]
USBIA0882-65C
- Antibody type:Primary
- Antigen name:adenylate kinase 1
- Antigen symbol:AK1
- Clonality:Monoclonal
- Clone:3A6-1F5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: AK1 (adenylate kinase 1)
Clonality: monoclonal
Clone: 3A6-1F5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human