Order Entry
Denmark
ContactUsLinkComponent
Anti-CREB3L4 Rabbit Polyclonal Antibody
Anti-CREB3L4 Rabbit Polyclonal Antibody
Catalog # USBIA0857-25A
Supplier:  US Biological
Anti-CREB3L4 Rabbit Polyclonal Antibody
Catalog # USBIA0857-25A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    cAMP responsive element binding Protein 3-like 4
  • Antigen symbol:
    CREB3L4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: CREB3L4 (cAMP responsive element binding protein 3-like 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human