Anti-Episialin, EMA Mouse Monoclonal Antibody [clone: EMA-39]
Catalog # USBI144491
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Episialin, EMA
- Clonality:Monoclonal
- Clone:EMA-39
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Format:ascites
- Form:lyophilized powder
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Episialin, EMA
Clonality: monoclonal
Clone: EMA-39
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human