Anti-TSPAN6 Rabbit Polyclonal Antibody
Catalog # USBI136501
Supplier: US Biological
Specifications
- Antistof type:Primary
- Antigen name:Tetraspanin 6
- Antigen symbol:TSPAN6
- Clonality:Polyclonal
- Konjugation:Unconjugated
- Vært:Rabbit
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Opbevaringstemperatur:Cool
- Koncentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Størrelse:50 µg
- Pakn:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: TSPAN6 (tetraspanin 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human