Order Entry
Denmark
ContactUsLinkComponent
Anti-PRKAB1 Rabbit Polyclonal Antibody
Anti-PRKAB1 Rabbit Polyclonal Antibody
Catalog # USBI135893
Supplier:  US Biological
Anti-PRKAB1 Rabbit Polyclonal Antibody
Catalog # USBI135893
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase AMP-Activated Non-Catalytic Subunit Beta 1
  • Antigen symbol:
    PRKAB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS,(without Mg2+, Ca2+), pH 7,4, 150mM sodium chloride, 0,02% sodium azide, 50% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: ELISA: 1:5000; Western Blot: 1:500-1:1000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: PRKAB1 (protein kinase, AMP-activated, beta 1 non-catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat