Anti-TBC1D8 Mouse Monoclonal Antibody [clone: 1A12]
Catalog # USBI134240
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:TBC1 domain family, member 8 (with GRAM domain)
- Antigen symbol:TBC1D8
- Clonality:Monoclonal
- Clone:1A12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 111
- Storage buffer:PBS, pH 7,2.
- Sequence:MEQLADVTLRRLLDNEVFDLDPDLQEPSQITKRDLEARAQNEFFRAFFRLPRKEKLHAVVDCSLWTPFSRCHTAGRMFASDSYICFASREDGCCKIILPLREVVSIEKME*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: TBC1D8 (TBC1 domain family, member 8 (with GRAM domain))
Clonality: monoclonal
Clone: 1A12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human