Anti-CYTH2 Mouse Monoclonal Antibody [clone: 6H5]
USBI131931
- Antibody type:Primary
- Antigen name:cytohesin 2
- Antigen symbol:CYTH2
- Clonality:Monoclonal
- Clone:6H5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat
- Western blot:Yes
- Form:liquid
- Amino acid number:314 to 398
- Storage buffer:PBS, pH 7,2.
- Sequence:VDDPRKPNCFELYIPNNKGQLIKACKTEADGRVVEGNHMVYRISAPTQEEKDEWIKSIQAAVSVDPFYEMLAARKKRISVKKKQE
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 3?g/ml
Type: Primary
Antigen: CYTH2 (cytohesin 2)
Clonality: monoclonal
Clone: 6H5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Rat