Order Entry
Denmark
ContactUsLinkComponent
Anti-PRKAB2 Mouse Monoclonal Antibody [clone: 2G9]
Anti-PRKAB2 Mouse Monoclonal Antibody [clone: 2G9]
Catalog # USBI131782
Supplier:  US Biological
Anti-PRKAB2 Mouse Monoclonal Antibody [clone: 2G9]
Catalog # USBI131782
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase AMP-Activated Non-Catalytic Subunit Beta 2
  • Antigen symbol:
    PRKAB2
  • Clonality:
    Monoclonal
  • Clone:
    2G9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2.
  • Sequence:
    MGNTTSDRVSGERHGAKAARSEGAGGHAPGKEHKIMVGSTDDPSVFSLPDSKLPGDKEFVSWQQDLEDSVKPTQQARPTVIRWSEGGKEVFISGSFNNWSTKIPLIKSHNDFVAILDLPEGEHQYKFFVDGQWVHDPSEPVVTSQLGTINNLIHVKKSDFEVFDALKLDSMESSETSCRDLSSSPPGPYGQEMYAFRSEERFKSPPILPPHLLQVILNKDTNISCDPALLPEPNHVMLNHLYALSIKDSVMVLSATHRYKKKYVTTLLYKPI
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: PRKAB2 (protein kinase, AMP-activated, beta 2 non-catalytic subunit)
Clonality: monoclonal
Clone: 2G9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human