Anti-PRKAA1 Mouse Polyclonal Antibody
Catalog # USBI131772
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Protein kinase, AMP-activated, alpha 1 catalytic subunit
- Antigen symbol:PRKAA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PRKAA1 (protein kinase, AMP-activated, alpha 1 catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human