Anti-MCC Mouse Monoclonal Antibody [clone: 1A7]
USBI129480
- Antibody type:Primary
- Antigen name:Mutated in colorectal cancers
- Antigen symbol:MCC
- Clonality:Monoclonal
- Clone:1A7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:DIVELNKRLQQTERERDLLEKKLAKAQCEQSHLMREHEDVQERTTLRYEERITELHSVIAELNKKIDRLQGTTIREEDEYSELRSELSQSQHEVNEDSRS
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: MCC (mutated in colorectal cancers)
Clonality: monoclonal
Clone: 1A7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human