Anti-Glutathione S-transferase A5, GST Class-alpha Member 5, Glutathione S-transferase A5-5 Rabbit Polyclonal Antibody
Catalog # USBI127371
Supplier: US Biological
Specifications
- Antibody type:Tag
- Antigen symbol:Glutathione S-transferase A5, GST Class-alpha Member 5, Glutathione S-transferase A5-5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MAEKPKLHYSNARGSMESIRWLLAAAGVELEEKFLESAEDLDKLRNDGSLLFQQVPMVEIDGMKLVQTRAILNYIASKYNLYGKDMKERALIDMYTEGIVDLTEMILLLLICQPEERDAKTALVKEKIKNRYFPAFEKVLKSHRQDYLVGNKLSWADIHLVELFYYVEELDSSLISSFPLLKALKTRISNLPTVKKFLQPGSQRKPPMDEKSLEEARKIFRF
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Tag
Antigen: Glutathione S-transferase A5, GST Class-alpha Member 5, Glutathione S-transferase A5-5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human