Anti-GAL3ST1 Rabbit Polyclonal Antibody
USBI127106
- Antibody type:Primary
- Antigen name:Galactose-3-O-sulfotransferase 1
- Antigen symbol:GAL3ST1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rabbit,Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MLPPQKKPWESMAKGLVLGALFTSFLLLMYSYAVPPLHAGLASTTPEAAASCSPPALEPEAVIRANGSAGECQPRRNIVFLKTHKTASSTLLNILFRFGQKHRLKFAFPNGRNDFDYPTFFARSLVQDYRPGACFNIICNHMRFHYDEVRGLVPTNAIFITVLRDPARLFESSFHYFGPVVPLTWKLSAGDKLTEFLQDPDRYYDPNGFNAHYLRNLLFFDLGYDNSLDPSSPQVQEHILEVERRFHLVLLQEYFDESLVLLKDLLCWELEDVLYFKLNARRDSPVPRLSGELYGRATAWNMLDSHLYRHFNASFWRKVEAFGRERMAREVAALRHANERMRTICIDGGHAVDAAAIQDEAMQPWQPLGTKSILGYNLKKSIGQRHAQLCRRMLTPEIQYLMDLGANLWVTKLWKFIRDFLRW
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: GAL3ST1 (galactose-3-O-sulfotransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rabbit