Order Entry
Denmark
Orders LinkContactUsLinkComponent
Anti-CTSC Rabbit Polyclonal Antibody

Anti-CTSC Rabbit Polyclonal Antibody

100 µg
  USBI124373
Anti-CTSC Rabbit Polyclonal Antibody
  USBI124373
Anti-CTSC Rabbit Polyclonal Antibody

100 µg

 
5.370,00 kr
   
1 * 1 Each
 

Type: Primary
Antigen: CTSC (cathepsin C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Documents

 
  • Size:
    100 µg
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    purified by Protein A chromatography
  • Antibody type:
    Primary
  • Form:
    liquid
  • Amino acid number:
    1 to 463
  • Isotype:
    IgG
  • Storage temperature:
    Cool
  • Host:
    Rabbit
  • Sequence:
    MGAGPSLLLAALLLLLSGDGAVRCDTPANCTYLDLLGTWVFQVGSSGSQRDVNCSVMGPQEKKVVVYLQKLDTAYDDLGNSGHFTIIYNQGFEIVLNDYKWFAFFKYKEEGSKVTTYCNETMTGWVHDVLGRNWACFTGKKVGTASENVYVNTAHLKNSQEKYSNRLYKYDHNFVKAINAIQKSWTATTYMEYETLTLGDMIRRSGGHSRKIPRPKPAPLTAEIQQKILHLPTSWDWRNVHGINFVSPVRNQASCGSCYSFASMGMLEARIRILTNNSQTPILSPQEVVSCSQYAQGCEGGFPYLIAGKYAQDFGLVEEACFPYTGTDSPCKMKEDCFRYYSSEYHYVGGFYGGCNEALMKLELVHHGPMAVAFEVYDDFLHYKKGIYHHTGLRDPFNPFELTNHAVLLVGYGTDSASGMDYWIVKNSWGTGWGENGYFRIRRGTDECAIESIAVAATPIPKL
  • Shipping temperature:
    Ice
  • Antigen name:
    cathepsin C
  • Cat. No.:
    USBI124373
  • Reactivity:
    Human
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    CTSC
  • Storage buffer:
    PBS, pH 7,2.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.