Anti-CAPN5 Rabbit Polyclonal Antibody
Catalog # USBI124324
Supplier: US Biological
Specifications
- Antistof type:Primary
- Antigen name:calpain 5
- Antigen symbol:CAPN5
- Clonality:Polyclonal
- Konjugation:Unconjugated
- Vært:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Aminosyretal:1 to 640
- Storage buffer:PBS, pH 7,2.
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Størrelse:100 µg
- Pakn:100 µG
Specifications
About this item
Type: Primary
Antigen: CAPN5 (calpain 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human