Anti-CRNN Rabbit Polyclonal Antibody
Catalog # USBI044682
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Cornulin
- Antigen symbol:CRNN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Form:liquid
- Amino acid number:259 to 274
- Storage buffer:PBS, 1% BSA, 0,01% sodium azide.
- Storage temperature:Cool
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CRNN (cornulin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: