Order Entry
Denmark
ContactUsLinkComponent
 
Anti-CYTH2 Rabbit Polyclonal Antibody (Biotin)
  USBI040539-BIOTIN
Anti-CYTH2 Rabbit Polyclonal Antibody (Biotin)
  USBI040539-BIOTIN

 

  • Antibody type:
    Primary
  • Antigen name:
    cytohesin 2
  • Antigen symbol:
    CYTH2
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Form:
    liquid
  • Amino acid number:
    35 to 64
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: CYTH2 (cytohesin 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse