Anti-PDE3B Rabbit Polyclonal Antibody
Catalog # USBI039801
Supplier: US Biological
Specifications
- Antistof type:Primary
- Antigen name:Phosphodiesterase 3B, cGMP-inhibited
- Antigen symbol:PDE3B
- Clonality:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Flowcytometri:Yes
- Vært:Rabbit
- ImmunFluorescens:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitop:internal
- Form:liquid
- Aminosyretal:407 to 435
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Størrelse:200 µl
- Pakn:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50
Type: Primary
Antigen: PDE3B (phosphodiesterase 3B, cGMP-inhibited)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human