Anti-NPY1R Rabbit Polyclonal Antibody (APC (Allophycocyanin))
USBI039106-APC
- Antibody type:Primary
- Antigen name:Neuropeptide Y receptor 1
- Antigen symbol:NPY1R
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: NPY1R (neuropeptide Y receptor Y1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human