Anti-MAP3K7 Rabbit Polyclonal Antibody (Biotin)
200 µl
Katalog # USBI038150-BIOTIN
Leverandør: US Biological
Anti-MAP3K7 Rabbit Polyclonal Antibody (Biotin)
Katalog # USBI038150-BIOTIN
Leverandør: US Biological
Gå til:
Product Details & Documents
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Type: Primary
Antigen: MAP3K7 (mitogen-activated protein kinase kinase kinase 7)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Documents
Specifikationer
- Size:200 µl
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by Protein A chromatography
- Antibody type:Primary
- Epitope:N-terminal
- Form:liquid
- Amino acid number:130 to 160
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Antigen name:Mitogen-activated protein kinase kinase kinase 7
- Cat. No.:USBI038150-BIOTIN
- Reactivity:Human|Mouse
- Conjugation:Biotin
- Antigen symbol:MAP3K7
- Storage buffer:PBS, pH 7,2
- ELISA:Yes
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



