Anti-IGF2AS Rabbit Polyclonal Antibody
200 µl
Catalog # USBI036908
Supplier: US Biological
Jump to:
Product Details & Documents
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: IGF2AS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Documents
Specifications
- Size:200 µl
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by Protein A chromatography
- Antibody type:Primary
- Epitope:internal
- Form:liquid
- Amino acid number:43 to 73
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Cat. No.:USBI036908
- Reactivity:Human
- Conjugation:Unconjugated
- Antigen symbol:IGF2AS
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



