Anti-FA49A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # USBI035320-APC
Supplier: US Biological
Specifications
- Antistof type:Primary
- Antigen symbol:FA49A
- Clonality:Polyclonal
- Konjugation:APC (Allophycocyanin)
- ELISA:Yes
- Flowcytometri:Yes
- Vært:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitop:N-terminal
- Form:liquid
- Aminosyretal:12 to 41
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Størrelse:200 µl
- Pakn:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: FA49A
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse