Ordreindtastning
Denmark
ContactUsLinkComponent
Anti-AVIL Rabbit Polyclonal Antibody
Anti-AVIL Rabbit Polyclonal Antibody
Katalog # USBI032332
Supplier:  US Biological
undefined
Anti-AVIL Rabbit Polyclonal Antibody
Katalog # USBI032332
Supplier:  US Biological

Specifications

  • Antistof type:
    Primary
  • Antigen name:
    Advillin
  • Antigen symbol:
    AVIL
  • Clonality:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flowcytometri:
    Yes
  • Vært:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitop:
    N-terminal
  • Form:
    liquid
  • Aminosyretal:
    183 to 212
  • Storage buffer:
    PBS, pH 7,2, 0,09% sodium azide.
  • Opbevaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Størrelse:
    200 µl
  • Pakn:
    200 µl

Specifications

About this item

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: AVIL (advillin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human