Anti-ACTA1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # USBI031573-APC
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Actin, alpha 1, skeletal muscle
- Antigen symbol:ACTA1
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Antigen synonyms:ASMA|ACTA
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000calc?l
Do not freeze!
Type: Primary
Antigen: ACTA1 (actin, alpha 1, skeletal muscle)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human