Ordreindtastning
Denmark
ContactUsLinkComponent
 
Anti-NTRK1 Mouse Monoclonal Antibody [clone: 185CT47.6.1]
  USBI031166
undefined
Anti-NTRK1 Mouse Monoclonal Antibody [clone: 185CT47.6.1]
  USBI031166

 

  • Antistof type:
    Primary
  • Antigen name:
    neurotrophic tyrosine kinase, receptor, type 1
  • Antigen symbol:
    NTRK1
  • Clonality:
    Monoclonal
  • Clone:
    185CT47.6.1
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Vært:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,09% sodium azide.
  • Opbevaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Størrelse:
    200 µl
  • Pakn:
    200 µl

 

 

Recommended dilution: Western Blot: 1:1000-1:500

Type: Primary
Antigen: NTRK1 (neurotrophic tyrosine kinase, receptor, type 1)
Clonality: Monoclonal
Clone: 185CT47.6.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse