Anti-ATP1A1 Chicken Polyclonal Antibody
ROCK200-9155
- Antibody type:Primary
- Antigen name:ATPase, Na+K+
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- ImmunoPrecipitation:Yes
- Reactivity:Dog
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: ATP1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Canine