Order Entry
Denmark
ContactUsLinkComponent
Anti-MYH (33-51) Rabbit Polyclonal Antibody
Anti-MYH (33-51) Rabbit Polyclonal Antibody
  PC709-100
Anti-MYH (33-51) Rabbit Polyclonal Antibody
  PC709-100

 

  • Antibody type:
    Primary
  • Antigen symbol:
    MYH
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: MYH (33-51)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: