Order Entry
Denmark
ContactUsLinkComponent
Anti-TP53 Rabbit Polyclonal Antibody
Anti-TP53 Rabbit Polyclonal Antibody
Catalog # PC386-50
Anti-TP53 Rabbit Polyclonal Antibody
Catalog # PC386-50

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tumor protein p53
  • Antigen symbol:
    P53
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: