Order Entry
Denmark
Orders LinkContactUsLinkComponent
Anti-NPY Rabbit Polyclonal Antibody

Anti-NPY Rabbit Polyclonal Antibody

100 µl
Catalog # PC223L-100
Anti-NPY Rabbit Polyclonal Antibody
Catalog # PC223L-100
Anti-NPY Rabbit Polyclonal Antibody

100 µl

Price
4.130,00 kr
Log in for availability
1 * 100 µl
Product Details & Documents

Type: Primary
Antigen: NPY (neuropeptide Y)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:

Specifications
  • Size:
    100 µl
  • Cat. No.:
    PC223L-100
  • Clonality:
    Polyclonal
  • Antibody type:
    Primary
  • Storage temperature:
    Cool
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    NPY
  • Shipping temperature:
    Ice
  • Antigen name:
    Neuropeptide Y
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.