Order Entry
Denmark
ContactUsLinkComponent
 
Anti-c-ErbB2/c-Neu (Ab-3) (3B5) Mouse Monoclonal Antibody
Catalog # OP15L-100
Anti-c-ErbB2/c-Neu (Ab-3) (3B5) Mouse Monoclonal Antibody
Catalog # OP15L-100

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    V-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2
  • Antigen symbol:
    C-ERBB2
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: c-ErbB2/c-Neu (Ab-3) (3B5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: