Order Entry
Denmark
ContactUsLinkComponent
Anti-CHUK Mouse Monoclonal Antibody [clone: 14A231]
Anti-CHUK Mouse Monoclonal Antibody [clone: 14A231]
Catalog # OP133-100
Anti-CHUK Mouse Monoclonal Antibody [clone: 14A231]
Catalog # OP133-100

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    conserved helix-loop-helix ubiquitous kinase
  • Antigen symbol:
    CHUK
  • Clonality:
    Monoclonal
  • Clone:
    14A231
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: CHUK (conserved helix-loop-helix ubiquitous kinase)
Clonality: monoclonal
Clone: 14A231
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: