Order Entry
Denmark
ContactUsLinkComponent
Anti-BRCA1 (Ab-4) (SD118) Mouse Monoclonal Antibody [clone: SD118]
Anti-BRCA1 (Ab-4) (SD118) Mouse Monoclonal Antibody [clone: SD118]
  OP107-100
Anti-BRCA1 (Ab-4) (SD118) Mouse Monoclonal Antibody [clone: SD118]
  OP107-100

 

  • Antibody type:
    Primary
  • Antigen symbol:
    BRCA1
  • Clonality:
    Monoclonal
  • Clone:
    SD118
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: BRCA1 (Ab-4) (SD118)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human