Order Entry
Denmark
ContactUsLinkComponent
Anti-UCHL1 Rabbit Polyclonal Antibody
Anti-UCHL1 Rabbit Polyclonal Antibody
Catalog # NE1013-50
Anti-UCHL1 Rabbit Polyclonal Antibody
Catalog # NE1013-50

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ubiquitin Carboxyl-Terminal Esterase L1
  • Antigen symbol:
    UCHL1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Amino acid number:
    176 to 191
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: UCHL1 (ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: