Anti-Glucocorticoid Receptor (Ab-2) (BuGR2) Mouse Monoclonal Antibody [clone: BuGR2]
GR32L-100
- Antibody type:Primary
- Antigen name:Glucocorticoid Receptor
- Antigen symbol:GCR
- Clonality:Monoclonal
- Clone:BuGR2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Glucocorticoid Receptor (Ab-2) (BuGR2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: